siRNA Details
Gene Symbol: LAMTOR5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102697.1 | 3 | 424 | |||
GenBank mRNA | BE547292.1 | - | 3 | 349 | ||
HY022359.1 | - | 3 | 414 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY005307.1 | - | 4 | 412 | Chr. 1 - 110407836 - 110416188 [+] on Build GRCh38 |
