siRNA Details
Gene Symbol: PCOLCE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038910.1 | 9 | 1955 | |||
GenBank mRNA | AK097705.1 | - | 9 | 1955 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PCOLCE-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038910.1 | 9 | 1955 | |||
GenBank mRNA | AK097705.1 | - | 9 | 1955 |