siRNA Details
Gene Symbol: TEX26-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038287.1 | 1 | 245 | |||
NR_038288.1 | 1 | 245 | ||||
GenBank mRNA | AK093165.1 | - | 1 | 245 | ||
BC042736.1 | - | 1 | 220 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TEX26-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038287.1 | 1 | 245 | |||
NR_038288.1 | 1 | 245 | ||||
GenBank mRNA | AK093165.1 | - | 1 | 245 | ||
BC042736.1 | - | 1 | 220 |