siRNA Details
Gene Symbol: POT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125718.1 | 10 | 1401 | |||
NR_125719.1 | 7 | 713 | ||||
GenBank mRNA | AK090675.1 | - | 4 | 346 | ||
BC142949.1 | - | 10 | 1357 | |||
BX648695.1 | - | 7 | 672 |
