siRNA Details
Gene Symbol: CASC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104154.1 | 1 | 80 | |||
GenBank mRNA | BC037927.2 | - | 1 | 80 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI544639.1 | - | 1 | 85 | Chr. 6 - 91629913 - 91690430 [-] on Build GRCh38 |
