siRNA Details
Gene Symbol: LOC101927839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110888.1 | 2 | 312 | |||
GenBank mRNA | BI461260.1 | - | 2 | 319 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI826307.1 | - | 2 | 308 | Chr. 17 - 2232685 - 2233593 [+] on Build GRCh38 |
