siRNA Details
Gene Symbol: FLJ46906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033896.1 | 4 | 669 | |||
GenBank mRNA | BC104460.2 | - | 3 | 650 | ||
BC104461.2 | - | 2 | 796 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ46906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033896.1 | 4 | 669 | |||
GenBank mRNA | BC104460.2 | - | 3 | 650 | ||
BC104461.2 | - | 2 | 796 |