siRNA Details
Gene Symbol: BOLA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045634.1 | 1 | 402 | |||
NR_045636.1 | 2 | 276 | ||||
NR_045637.1 | 1 | 402 | ||||
GenBank mRNA | AK123453.1 | - | 2 | 212 | ||
BC030102.2 | - | 1 | 347 | |||
DA356576.1 | - | 1 | 374 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG706622.1 | - | 1 | 352 | Chr. 2 - 74148023 - 74149056 [+] on Build GRCh38 |
