siRNA Details
Gene Symbol: LOC101927406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110222.1 | 3 | 367 | |||
GenBank mRNA | BC038548.1 | - | 3 | 354 | ||
DA512861.1 | - | 3 | 367 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101927406 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110222.1 | 3 | 367 | |||
GenBank mRNA | BC038548.1 | - | 3 | 354 | ||
DA512861.1 | - | 3 | 367 |