siRNA Details
Gene Symbol: LOC101927523 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120624.1 | 1, 2 | 140 | |||
GenBank mRNA | BC039476.1 | - | 1, 2 | 137 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM453629.1 | - | 1, 2 | 136 | Chr. 10 - 104475078 - 104480274 [-] on Build GRCh38 |
