siRNA Details
Gene Symbol: PROSER2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038222.1 | 6 | 1929 | |||
GenBank mRNA | AK056703.1 | - | 6 | 1918 | ||
BC032757.1 | - | 1 | 347 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PROSER2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038222.1 | 6 | 1929 | |||
GenBank mRNA | AK056703.1 | - | 6 | 1918 | ||
BC032757.1 | - | 1 | 347 |