siRNA Details
Gene Symbol: TRAM2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103446.1 | 1 | 971 | |||
NR_103447.1 | 2 | 477 | ||||
GenBank mRNA | AK095117.1 | - | 1 | 866 | ||
BE741421.1 | - | 2 | 289 | |||
DB479639.1 | - | 2 | 479 |
