siRNA Details
Gene Symbol: RUSC1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039517.1 | NP_001034606.1 | 2 | 193 | ||
GenBank mRNA | BC062571.1 | - | 2 | 222 | 5183308 | |
BC080538.1 | - | 2 | 193 | 5018492 |
