siRNA Details
Gene Symbol: UBXN10-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110078.1 | 1 | 114 | |||
GenBank mRNA | BX106299.1 | - | 1 | 101 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA479461.1 | - | 1 | 98 | Chr. 1 - 20184378 - 20186473 [-] on Build GRCh38 |
