siRNA Details
Gene Symbol: PRICKLE2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046702.1 | 4 | 1529 | |||
GenBank mRNA | AK056491.1 | - | 4 | 1529 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PRICKLE2-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046702.1 | 4 | 1529 | |||
GenBank mRNA | AK056491.1 | - | 4 | 1529 |