siRNA Details
Gene Symbol: LINC01040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131228.1 | 1 | 69 | |||
GenBank mRNA | BG192883.1 | - | 1 | 65 | ||
BG203629.1 | - | 1 | 67 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131228.1 | 1 | 69 | |||
GenBank mRNA | BG192883.1 | - | 1 | 65 | ||
BG203629.1 | - | 1 | 67 |