siRNA Details
Gene Symbol: LINC01040 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131228.1 | 2 | 525 | |||
GenBank mRNA | BG192883.1 | - | 2 | 521 | ||
BG203629.1 | - | 2 | 523 | |||
BG205080.1 | - | 1 | 392 | |||
BG212965.1 | - | 1 | 392 |
