siRNA Details
Gene Symbol: ZNF503-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038223.1 | 3 | 902 | |||
NR_038224.1 | 3 | 560 | ||||
NR_038225.1 | 4 | 671 | ||||
NR_038226.1 | 4 | 683 | ||||
GenBank mRNA | AK023445.1 | - | 3 | 895 | ||
CB997793.1 | - | 3 | 588 | |||
CN360505.1 | - | 4 | 569 |
