siRNA Details
Gene Symbol: SLC39A12-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038419.1 | 1 | 18 | |||
GenBank mRNA | AK055012.1 | - | 1 | 18 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC39A12-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038419.1 | 1 | 18 | |||
GenBank mRNA | AK055012.1 | - | 1 | 18 |