siRNA Details
Gene Symbol: BAIAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026857.1 | 2 | 1872 | |||
GenBank mRNA | AK056555.1 | - | 2 | 1854 | ||
AK096609.1 | - | 1 | 780 | |||
AK124762.1 | - | 2 | 1913 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BAIAP2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026857.1 | 2 | 1872 | |||
GenBank mRNA | AK056555.1 | - | 2 | 1854 | ||
AK096609.1 | - | 1 | 780 | |||
AK124762.1 | - | 2 | 1913 |