siRNA Details
Gene Symbol: GAS1RR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049794.1 | 3 | 525 | |||
GenBank mRNA | BC036850.1 | - | 3 | 536 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GAS1RR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_049794.1 | 3 | 525 | |||
GenBank mRNA | BC036850.1 | - | 3 | 536 |