siRNA Details
Gene Symbol: ISM1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040043.1 | 3 | 388 | |||
GenBank mRNA | BC038745.1 | - | 3 | 375 | ||
BX092222.1 | - | 1 | 26 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ISM1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040043.1 | 3 | 388 | |||
GenBank mRNA | BC038745.1 | - | 3 | 375 | ||
BX092222.1 | - | 1 | 26 |