siRNA Details
Gene Symbol: ARHGAP26-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046680.1 | 2 | 149 | |||
GenBank mRNA | BC039527.1 | - | 2 | 149 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM564058.1 | - | 2 | 167 | Chr. 5 - 142859602 - 142868915 [-] on Build GRCh38 |
