siRNA Details
Gene Symbol: NUP50-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038956.1 | 3 | 784 | |||
NR_038957.1 | 3 | 759 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX389200.2 | - | 1 | 81 | Chr. 22 - 45133776 - 45134371 [-] on Build GRCh38 |
