siRNA Details
Gene Symbol: SLC16A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BX648855.1 | - | 3 | 1408 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY180165.1 | - | 3 | 488 | Chr. 1 - 112956616 - 112968099 [+] on Build GRCh38 |
