siRNA Details
Gene Symbol: UPK1A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046420.1 | 2 | 261 | |||
GenBank mRNA | BC007817.1 | - | 2 | 270 | ||
BF683645.1 | - | 2 | 262 | |||
BX283291.1 | - | 2 | 262 | |||
EL733629.1 | - | 2 | 221 |
