siRNA Details
Gene Symbol: UPK1A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046420.1 | 1 | 31 | |||
GenBank mRNA | BC007817.1 | - | 1 | 40 | ||
BF683645.1 | - | 1 | 32 | |||
BX283291.1 | - | 1 | 32 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UPK1A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046420.1 | 1 | 31 | |||
GenBank mRNA | BC007817.1 | - | 1 | 40 | ||
BF683645.1 | - | 1 | 32 | |||
BX283291.1 | - | 1 | 32 |