siRNA Details
Gene Symbol: TEX41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033870.2 | 5 | 761 | |||
GenBank mRNA | BC043549.1 | - | 5 | 702 | ||
BX648102.1 | - | 4 | 606 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TEX41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033870.2 | 5 | 761 | |||
GenBank mRNA | BC043549.1 | - | 5 | 702 | ||
BX648102.1 | - | 4 | 606 |