siRNA Details
Gene Symbol: LOC101928525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121579.1 | 2, 3 | 686 | |||
GenBank mRNA | BC015688.1 | - | 2, 3 | 686 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121579.1 | 2, 3 | 686 | |||
GenBank mRNA | BC015688.1 | - | 2, 3 | 686 |