siRNA Details
Gene Symbol: ITGB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038311.1 | 1 | 184 | |||
NR_038312.1 | 1 | 184 | ||||
NR_038313.1 | 1 | 184 | ||||
NR_038314.1 | 1 | 184 | ||||
NR_038315.1 | 1 | 184 | ||||
NR_038316.1 | 1 | 184 | ||||
GenBank mRNA | BC040064.1 | - | 1 | 168 | ||
DA378472.1 | - | 1 | 185 | |||
DA942354.1 | - | 1 | 147 | |||
DA944688.1 | - | 1 | 146 | |||
DA948979.1 | - | 1 | 149 | |||
DB115909.1 | - | 1 | 146 |
