siRNA Details
Gene Symbol: ITGB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038311.1 | 5 | 1041 | |||
NR_038312.1 | 4 | 998 | ||||
NR_038313.1 | 5 | 1017 | ||||
NR_038314.1 | 4 | 946 | ||||
NR_038315.1 | 4 | 942 | ||||
NR_038316.1 | 3 | 903 | ||||
GenBank mRNA | BC040064.1 | - | 4 | 930 |
