siRNA Details
Gene Symbol: LBX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029380.1 | 2 | 266 | |||
GenBank mRNA | AK123344.1 | - | 2 | 266 | ||
AK294034.1 | - | 2 | 159 | |||
LC015092.1 | - | 2 | 284 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LBX1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029380.1 | 2 | 266 | |||
GenBank mRNA | AK123344.1 | - | 2 | 266 | ||
AK294034.1 | - | 2 | 159 | |||
LC015092.1 | - | 2 | 284 |