siRNA Details
Gene Symbol: LOC101928418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104605.1 | 3 | 1113 | |||
GenBank mRNA | BC030770.2 | - | 3 | 1113 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928418 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104605.1 | 3 | 1113 | |||
GenBank mRNA | BC030770.2 | - | 3 | 1113 |