siRNA Details
Gene Symbol: DIAPH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046539.2 | 2, 3 | 234 | |||
NR_051994.2 | 2, 3 | 162 | ||||
GenBank mRNA | BX093461.1 | - | 2, 3 | 233 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DIAPH3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046539.2 | 2, 3 | 234 | |||
NR_051994.2 | 2, 3 | 162 | ||||
GenBank mRNA | BX093461.1 | - | 2, 3 | 233 |