siRNA Details
Gene Symbol: SLC2A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033967.1 | 1 | 276 | |||
GenBank mRNA | AK056786.1 | - | 1 | 221 | ||
BU657382.1 | - | 1 | 283 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLC2A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033967.1 | 1 | 276 | |||
GenBank mRNA | AK056786.1 | - | 1 | 221 | ||
BU657382.1 | - | 1 | 283 |