siRNA Details
Gene Symbol: SPAG5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040012.1 | 3 | 335 | |||
GenBank mRNA | AK098638.1 | - | 3 | 523 | ||
AL157421.1 | - | 1 | 664 | |||
BC001556.1 | - | 2 | 1113 | |||
DB457167.1 | - | 3 | 312 |
