siRNA Details
Gene Symbol: LOC100506725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108082.1 | 1 | 380 | |||
NR_108083.1 | 1 | 380 | ||||
GenBank mRNA | DT218025.1 | - | 1 | 371 | ||
HY127409.1 | - | 1 | 90 | |||
HY153176.1 | - | 1 | 381 |
