siRNA Details
Gene Symbol: CERS6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045786.1 | 4 | 2613 | |||
NR_045787.1 | 3 | 2520 | ||||
GenBank mRNA | BC035245.1 | - | 3 | 2520 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CERS6-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045786.1 | 4 | 2613 | |||
NR_045787.1 | 3 | 2520 | ||||
GenBank mRNA | BC035245.1 | - | 3 | 2520 |