siRNA Details
Gene Symbol: FAM212B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038951.1 | 3 | 2037 | |||
NR_038952.1 | 2 | 1882 | ||||
GenBank mRNA | AK092511.1 | - | 3 | 1923 | ||
BC041428.1 | - | 3 | 2037 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM212B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038951.1 | 3 | 2037 | |||
NR_038952.1 | 2 | 1882 | ||||
GenBank mRNA | AK092511.1 | - | 3 | 1923 | ||
BC041428.1 | - | 3 | 2037 |