siRNA Details
Gene Symbol: FAM212B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038951.1 | 3 | 684 | |||
NR_038952.1 | 2 | 529 | ||||
GenBank mRNA | AK092511.1 | - | 3 | 570 | ||
BC041428.1 | - | 3 | 684 | |||
BG329450.1 | - | 2 | 516 |
