siRNA Details
Gene Symbol: FLJ31104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102755.1 | 2 | 625 | |||
GenBank mRNA | AK055666.1 | - | 2 | 472 | ||
BC039324.1 | - | 2 | 625 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FLJ31104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_102755.1 | 2 | 625 | |||
GenBank mRNA | AK055666.1 | - | 2 | 472 | ||
BC039324.1 | - | 2 | 625 |