siRNA Details
Gene Symbol: LOC105371485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135642.1 | 2 | 1356 | |||
GenBank mRNA | BC040634.1 | - | 2 | 1189 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC105371485 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135642.1 | 2 | 1356 | |||
GenBank mRNA | BC040634.1 | - | 2 | 1189 |