siRNA Details
Gene Symbol: LOC286238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100111.1 | NP_001093581.1 | 2 | 324 | ||
GenBank mRNA | AK098532.1 | - | 2 | 302 | ||
BC101432.1 | - | 2 | 254 | |||
BC101439.1 | - | 2 | 254 | |||
BC101440.1 | - | 2 | 254 | |||
BC101442.1 | - | 2 | 254 | |||
DB454766.2 | - | 2 | 324 |
