siRNA Details
Gene Symbol: LOC286238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100111.1 | NP_001093581.1 | 1 | 50 | ||
GenBank mRNA | AK098532.1 | - | 1 | 28 | ||
DB454766.2 | - | 1 | 50 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC286238 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100111.1 | NP_001093581.1 | 1 | 50 | ||
GenBank mRNA | AK098532.1 | - | 1 | 28 | ||
DB454766.2 | - | 1 | 50 |