siRNA Details
Gene Symbol: ITPKB-IT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103784.1 | 2 | 170 | |||
GenBank mRNA | BI116225.1 | - | 2 | 101 | ||
BI906918.1 | - | 2 | 169 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR744533.1 | - | 2 | 67 | Chr. 1 - 226656642 - 226674965 [-] on Build GRCh38 |
