siRNA Details
Gene Symbol: LINC01185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033980.1 | 3 | 708 | |||
GenBank mRNA | AK122786.1 | - | 3 | 708 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01185 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033980.1 | 3 | 708 | |||
GenBank mRNA | AK122786.1 | - | 3 | 708 |