siRNA Details
Gene Symbol: TSTD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010141.1 | XP_016865630.1 | 1 | 273 |
Gene Symbol: LOC101927365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK025621.1 | - | 1 | 166 | ||
BC029846.1 | - | 1 | 205 | |||
BC047485.1 | - | 1 | 205 |
