siRNA Details
Gene Symbol: CCDC183-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024580.1 | 1 | 42 | |||
GenBank mRNA | AK055547.1 | - | 1 | 42 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA573712.1 | - | 1 | 42 | Chr. 9 - 136806486 - 136808848 [-] on Build GRCh38 |
