siRNA Details
Gene Symbol: TNKS2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108070.1 | 1 | 338 | |||
GenBank mRNA | BM678746.1 | - | 1 | 338 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BP396244.1 | - | 1 | 101 | Chr. 10 - 91782844 - 91797959 [-] on Build GRCh38 |
