siRNA Details
Gene Symbol: UBAC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036531.1 | 2 | 1574 | |||
NR_036532.1 | 2 | 1300 | ||||
GenBank mRNA | AF339765.1 | - | 1 | 772 | ||
AK097488.1 | - | 2 | 1579 | |||
BC043515.1 | - | 2 | 1255 |
